missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-[(1-phenyl-1H-imidazol-2-yl)methyl]amine, Thermo Scientific™

Catalog No. p-4391273
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC75946CB 250mg Amber Glass Bottle
CC75946DA 1g Amber Glass Bottle
CC75946DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC75946CB Supplier Thermo Scientific Chemicals Supplier No. CC75946CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1034566-07-8
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.25
InChI Key IZYNHWOVGJNYQF-UHFFFAOYSA-N
Synonym methyl 1-phenylimidazol-2-yl methyl amine, n-methyl-1-phenyl-1h-imidazol-2-yl methyl amine, n-methyl-1-1-phenyl-1h-imidazol-2-yl methanamine
PubChem CID 45594302

Specifications

Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material N-methyl-[(1-phenyl-1H-imidazol-2-yl)methyl]amine
Hazard Category GHS Signal Word: Danger