missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-methyl-[(1-phenyl-1H-imidazol-2-yl)methyl]amine, Thermo Scientific™
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Quantity | 5g |
| Formula Weight | 187.24 |
| Percent Purity | 97% |
| Chemical Name or Material | N-methyl-[(1-phenyl-1H-imidazol-2-yl)methyl]amine |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |