missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-1-naphthylmethylamine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America M098425G
Specifications
| N-Methyl-1-naphthylmethylamine Hydrochloride | |
| 193°C | |
| C12H14ClN | |
| 25 g | |
| BVJVHPKFDIYQOU-UHFFFAOYSA-N | |
| N-methyl-1-naphthalen-1-ylmethanamine;hydrochloride | |
| 16211748 | |
| ≥98.0% (T) |
| 65473-13-4 | |
| White-Yellow | |
| MFCD00012555 | |
| n-methyl-1-naphthalenemethylamine hydrochloride, n-methyl-1-naphthalen-1-yl methanamine hydrochloride, n-methyl-1-naphthylmethylamine hydrochloride, n-methyl-n-naphthylmethylamine hydrochloride, unii-z8vrt9b309, methyl naphthalen-1-ylmethyl amine hydrochloride, n-methyl-1-naphthalenemethanamine hydrochloride, n-methyl-c-naphthalen-1-yl methanamine hydrochloride, 1-naphthalenemethanamine, n-methyl-, hydrochloride 1:1, 1-methylaminomethyl naphthalene hydrochloride | |
| CNCC1=CC=CC2=CC=CC=C21.Cl | |
| 207.701 | |
| 207.70 | |
| Crystalline Powder |
Chemical Identifiers
| 65473-13-4 | |
| 207.701 | |
| BVJVHPKFDIYQOU-UHFFFAOYSA-N | |
| 16211748 | |
| CNCC1=CC=CC2=CC=CC=C21.Cl |
| C12H14ClN | |
| MFCD00012555 | |
| n-methyl-1-naphthalenemethylamine hydrochloride, n-methyl-1-naphthalen-1-yl methanamine hydrochloride, n-methyl-1-naphthylmethylamine hydrochloride, n-methyl-n-naphthylmethylamine hydrochloride, unii-z8vrt9b309, methyl naphthalen-1-ylmethyl amine hydrochloride, n-methyl-1-naphthalenemethanamine hydrochloride, n-methyl-c-naphthalen-1-yl methanamine hydrochloride, 1-naphthalenemethanamine, n-methyl-, hydrochloride 1:1, 1-methylaminomethyl naphthalene hydrochloride | |
| N-methyl-1-naphthalen-1-ylmethanamine;hydrochloride |
Safety and Handling
TSCA : No