missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1-naphthylamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7132575
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11725G 5 g
M117225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11725G Supplier TCI America Supplier No. M11725G

Chemical Identifiers

CAS 4643-36-1
Molecular Formula C11H12ClN
Molecular Weight (g/mol) 193.674
MDL Number MFCD00191555
InChI Key ALZGTEASAJRPTL-UHFFFAOYSA-N
Synonym 1-(N-Methylamino)naphthalene Hydrochloride
PubChem CID 21191510
IUPAC Name N-methylnaphthalen-1-amine;hydrochloride
SMILES CNC1=CC=CC2=CC=CC=C21.Cl

Specifications

Melting Point 177°C
Quantity 5 g
Formula Weight 193.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-1-naphthylamine Hydrochloride