missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1-naphthalenemethylamine hydrochloride, 98%

Catalog No. AAH2871903
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2871903 1 g
AAH2871909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2871903 Supplier Thermo Scientific Chemicals Supplier No. H2871903

CAS: 65473-13-4 | C12H14ClN | 207.701 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65473-13-4
Molecular Formula C12H14ClN
Molecular Weight (g/mol) 207.701
MDL Number MFCD00012555
InChI Key BVJVHPKFDIYQOU-UHFFFAOYSA-N
Synonym n-methyl-1-naphthalenemethylamine hydrochloride, n-methyl-1-naphthalen-1-yl methanamine hydrochloride, n-methyl-1-naphthylmethylamine hydrochloride, n-methyl-n-naphthylmethylamine hydrochloride, unii-z8vrt9b309, methyl naphthalen-1-ylmethyl amine hydrochloride, n-methyl-1-naphthalenemethanamine hydrochloride, n-methyl-c-naphthalen-1-yl methanamine hydrochloride, 1-naphthalenemethanamine, n-methyl-, hydrochloride 1:1, 1-methylaminomethyl naphthalene hydrochloride
PubChem CID 16211748
IUPAC Name N-methyl-1-naphthalen-1-ylmethanamine;hydrochloride
SMILES CNCC1=CC=CC2=CC=CC=C21.Cl

Specifications

Melting Point 190°C to 193°C
Quantity 1 g
Formula Weight 207.7
Percent Purity 98%
Chemical Name or Material N-Methyl-1-naphthalenemethylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only