missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(1-methylindolin-5-yl)methylamine, 90%, Thermo Scientific™

Chemical Identifiers

CAS 906352-81-6
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD09817499
InChI Key QPUPWFNEDMKWGL-UHFFFAOYSA-N
Synonym 1-methyl-5-methylamino methyl indoline, 2,3-dihydro-1-methyl-5-methylamino methyl-1h-indole, methyl 1-methyl-2,3-dihydroindol-5-yl methyl amine, methyl 1-methyl-2,3-dihydro-1h-indol-5-yl methyl amine, methyl 1-methylindolin-5-yl methyl amine, n-methyl-1-1-methylindolin-5-yl methanamine, 1-2,3-dihydro-1-methyl-1h-indol-5-yl-n-methylmethylamine, n-methyl-1-1-methyl-2,3-dihydro-1h-indol-5-yl methanamine
PubChem CID 24229592
IUPAC Name N-methyl-1-(1-methyl-2,3-dihydroindol-5-yl)methanamine
SMILES CNCC1=CC2=C(C=C1)N(C)CC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC58746CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC58746DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

906352-81-6
176.26
QPUPWFNEDMKWGL-UHFFFAOYSA-N
24229592
CNCC1=CC2=C(C=C1)N(C)CC2
C11H16N2
MFCD09817499
1-methyl-5-methylamino methyl indoline, 2,3-dihydro-1-methyl-5-methylamino methyl-1h-indole, methyl 1-methyl-2,3-dihydroindol-5-yl methyl amine, methyl 1-methyl-2,3-dihydro-1h-indol-5-yl methyl amine, methyl 1-methylindolin-5-yl methyl amine, n-methyl-1-1-methylindolin-5-yl methanamine, 1-2,3-dihydro-1-methyl-1h-indol-5-yl-n-methylmethylamine, n-methyl-1-1-methyl-2,3-dihydro-1h-indol-5-yl methanamine
N-methyl-1-(1-methyl-2,3-dihydroindol-5-yl)methanamine

Specifications

906352-81-6
100.0
C11H16N2
1-methyl-5-methylamino methyl indoline, 2,3-dihydro-1-methyl-5-methylamino methyl-1h-indole, methyl 1-methyl-2,3-dihydroindol-5-yl methyl amine, methyl 1-methyl-2,3-dihydro-1h-indol-5-yl methyl amine, methyl 1-methylindolin-5-yl methyl amine, n-methyl-1-1-methylindolin-5-yl methanamine, 1-2,3-dihydro-1-methyl-1h-indol-5-yl-n-methylmethylamine, n-methyl-1-1-methyl-2,3-dihydro-1h-indol-5-yl methanamine
CNCC1=CC2=C(C=C1)N(C)CC2
176.26
176.26
N-methyl-(1-methylindolin-5-yl)methylamine
89.5
90%
MFCD09817499
QPUPWFNEDMKWGL-UHFFFAOYSA-N
N-methyl-1-(1-methyl-2,3-dihydroindol-5-yl)methanamine
24229592
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents