missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methylamine, 90%, Thermo Scientific™

Chemical Identifiers

CAS 912569-73-4
Molecular Formula C13H14F3N3O
Molecular Weight (g/mol) 285.27
MDL Number MFCD09817546
InChI Key AJQKRVBDVZVOBK-UHFFFAOYSA-N
Synonym methyl 1-methyl-5-phenoxy-3-trifluoromethyl pyrazol-4-yl methyl amine, n-methyl-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methylamine, n-methyl-1-methyl-5-phenoxy-3-1h-pyrazol-4-yl methylamine, n-methyl-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methylamine,97+%, n-methyl-1-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methanamine
PubChem CID 24229724
IUPAC Name N-methyl-1-[1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazol-4-yl]methanamine
SMILES CNCC1=C(N(N=C1C(F)(F)F)C)OC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC64846CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC64846DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

912569-73-4
285.27
AJQKRVBDVZVOBK-UHFFFAOYSA-N
24229724
CNCC1=C(N(N=C1C(F)(F)F)C)OC2=CC=CC=C2
C13H14F3N3O
MFCD09817546
methyl 1-methyl-5-phenoxy-3-trifluoromethyl pyrazol-4-yl methyl amine, n-methyl-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methylamine, n-methyl-1-methyl-5-phenoxy-3-1h-pyrazol-4-yl methylamine, n-methyl-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methylamine,97+%, n-methyl-1-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methanamine
N-methyl-1-[1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazol-4-yl]methanamine

Specifications

912569-73-4
100.0
C13H14F3N3O
methyl 1-methyl-5-phenoxy-3-trifluoromethyl pyrazol-4-yl methyl amine, n-methyl-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methylamine, n-methyl-1-methyl-5-phenoxy-3-1h-pyrazol-4-yl methylamine, n-methyl-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methylamine,97+%, n-methyl-1-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methanamine
CNCC1=C(N(N=C1C(F)(F)F)C)OC2=CC=CC=C2
285.27
285.27
N-methyl-[1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methylamine
89.5
90%
MFCD09817546
AJQKRVBDVZVOBK-UHFFFAOYSA-N
N-methyl-1-[1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazol-4-yl]methanamine
24229724
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents