missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[1-(6-methylpyrazin-2-yl)piperid-4-yl]methylamine, 90%, Thermo Scientific™

Chemical Identifiers

CAS 887922-91-0
Molecular Formula C12H20N4
MDL Number MFCD09064987
Synonym n-methyl-1-6-methylpyrazin-2-yl piperid-4-yl methylamine, methyl 1-6-methylpyrazin-2-yl piperidin-4-yl methyl amine, methyl 1-6-methylpyrazin-2-yl 4-piperidyl methyl amine, n-methyl-1-1-6-methylpyrazin-2-yl piperidin-4-yl methanamine
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC58946CB
SDS View Documents
Thermo Scientific Chemicals
CC58946CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC58946DA
SDS View Documents
Thermo Scientific Chemicals
CC58946DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

887922-91-0
C12H20N4
MFCD09064987
n-methyl-1-6-methylpyrazin-2-yl piperid-4-yl methylamine, methyl 1-6-methylpyrazin-2-yl piperidin-4-yl methyl amine, methyl 1-6-methylpyrazin-2-yl 4-piperidyl methyl amine, n-methyl-1-1-6-methylpyrazin-2-yl piperidin-4-yl methanamine

Specifications

887922-91-0
100.0
Amber Glass Bottle
MFCD09064987
n-methyl-1-6-methylpyrazin-2-yl piperid-4-yl methylamine, methyl 1-6-methylpyrazin-2-yl piperidin-4-yl methyl amine, methyl 1-6-methylpyrazin-2-yl 4-piperidyl methyl amine, n-methyl-1-1-6-methylpyrazin-2-yl piperidin-4-yl methanamine
90%
89.5
90%
C12H20N4
250mg
220.32
N-methyl-[1-(6-methylpyrazin-2-yl)piperid-4-yl]methylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents