missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1,3-diaminopropane 98.0+%, TCI America™

Catalog No. p-7132258
Change view
Click to view available options
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
M059925ML 25 mL
M0599100ML 100 mL
M0599500ML 500 mL
3 options

Catalog No. M059925ML

Supplier: TCI America M059925ML

Only null left

Chemical Identifiers

CAS 6291-84-5
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008209
InChI Key QHJABUZHRJTCAR-UHFFFAOYSA-N
Synonym n-methyl-1,3-propanediamine, n-methyl-1,3-diaminopropane, 3-methylamino propylamine, 1,3-propanediamine, n-methyl, n-methylpropane-1,3-diamine, 3-aminopropyl methylamine, n-methyltrimethylenediamine, 3-aminopropylmethylamine, n-methyl-1,3-propylenediamine, 1-amino-3-methylamino propane
PubChem CID 80511
IUPAC Name N'-methylpropane-1,3-diamine
SMILES CNCCCN

Specifications

Color Colorless
Boiling Point 143°C
Quantity 25 mL
UN Number 2734
Formula Weight 88.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyl-1,3-diaminopropane
EINECSNumber (9)-1389
RTECSNumber TX8242500
TSCA Yes