missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1,3-diaminopropane 98.0+%, TCI America™

Catalog No. m0599100ml
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0599100ML 100 mL
M059925ML 25 mL
M0599500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0599100ML Supplier TCI America Supplier No. M0599100ML
Only null left

Chemical Identifiers

CAS 6291-84-5
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008209
InChI Key QHJABUZHRJTCAR-UHFFFAOYSA-N
Synonym n-methyl-1,3-propanediamine, n-methyl-1,3-diaminopropane, 3-methylamino propylamine, 1,3-propanediamine, n-methyl, n-methylpropane-1,3-diamine, 3-aminopropyl methylamine, n-methyltrimethylenediamine, 3-aminopropylmethylamine, n-methyl-1,3-propylenediamine, 1-amino-3-methylamino propane
PubChem CID 80511
IUPAC Name N'-methylpropane-1,3-diamine
SMILES CNCCCN

Specifications

Color Colorless
Boiling Point 143°C
Quantity 100 mL
UN Number 2734
Formula Weight 88.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyl-1,3-diaminopropane
EINECSNumber (9)-1389
RTECSNumber TX8242500
TSCA Yes