missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1,2-phenylenediamine, 97%

Catalog No. AC434230050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434230050 5 mL Glass bottle
AC434230250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434230050 Supplier Thermo Scientific Chemicals Supplier No. 434230050

CAS: 4760-34-3 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4760-34-3
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00192314
InChI Key RPKCLSMBVQLWIN-UHFFFAOYSA-N
Synonym n1-methylbenzene-1,2-diamine, n-methyl-o-phenylenediamine, n-methylbenzene-1,2-diamine, 1,2-benzenediamine, n-methyl, n-methyl-1,2-phenylenediamine, n-methyl-1,2-benzenediamine, 2-amino-n-methylaniline, n-methyl-2-phenylenediamine, 2-methylamino aniline, unii-roi30fpr50
PubChem CID 78498
IUPAC Name 2-N-methylbenzene-1,2-diamine
SMILES CNC1=CC=CC=C1N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 22°C
Color Brown to Yellow
Density 1.0750g/mL
Boiling Point 123°C to 124°C (10.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 mL
Specific Gravity 1.075
Formula Weight 122.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Methyl-1, 2-phenylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-297-6

RUO – Research Use Only