missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylcarbamoyl Chloride 98.0+%, TCI America™

Catalog No. p-7133033
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21171G 1 g
M21175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21171G Supplier TCI America Supplier No. M21171G

Chemical Identifiers

CAS 30289-28-2
Molecular Formula C3H6ClNO2
Molecular Weight (g/mol) 123.54
MDL Number MFCD06658962
InChI Key SHQJHXLWZDMQEJ-UHFFFAOYSA-N
Synonym acmc-209he9, methylmethoxycarbamoyl chloride, methoxy methyl carbamic chloride, n-methoxy-n-methylcarbamic chloride, n-methoxy-n-methylcarbamyl chloride, n-methyl-n-methoxycarbamyl chloride, carbamic chloride,n-methoxy-n-methyl, n-methoxy-n-methylcarbamic acid chloride
PubChem CID 2734926
IUPAC Name N-methoxy-N-methylcarbamoyl chloride
SMILES CON(C)C(Cl)=O

Specifications

Color Yellow
Boiling Point 60°C
Quantity 1 g
UN Number 3265
Formula Weight 123.54
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methoxy-N-methylcarbamoyl Chloride