Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Methoxy-N-methylcarbamoyl Chloride 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 60°C |
| Quantity | 1 g |
| UN Number | 3265 |
| Formula Weight | 123.54 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | N-Methoxy-N-methylcarbamoyl Chloride |
Safety and Handling
| TSCA | No |