missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylacetamide 98.0+%, TCI America™

Catalog No. p-7132443
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M091910G 10 g
M091925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M091910G Supplier TCI America Supplier No. M091910G
Only null left

Chemical Identifiers

CAS 78191-00-1
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00060098
InChI Key OYVXVLSZQHSNDK-UHFFFAOYSA-N
Synonym n-methyl-n-methoxyacetamide, acetamide, n-methoxy-n-methyl, n-methoxyl-n-methylacetamide, n-methoxy-n-methyl-acetamide, n-methoxy-n-methylacetamid, pubchem12775, acmc-209pdo, n-methoxy-n-methylacetamde, n-methoxy n-methylacetamide, n-methoxy-n-methyl acetamide
PubChem CID 537505
IUPAC Name N-methoxy-N-methylacetamide
SMILES CON(C)C(C)=O

Specifications

Color Colorless
Boiling Point 94°C
Quantity 10 g
UN Number 1993
Formula Weight 103.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methoxy-N-methylacetamide