missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylacetamide, 98%

Catalog No. p-2691943
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC369310050 5 g Glass bottle
AC369310250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC369310050 Supplier Thermo Scientific Chemicals Supplier No. 369310050

CAS: 78191-00-1 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78191-00-1
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00060098
InChI Key OYVXVLSZQHSNDK-UHFFFAOYSA-N
Synonym n-methyl-n-methoxyacetamide, acetamide, n-methoxy-n-methyl, n-methoxyl-n-methylacetamide, n-methoxy-n-methyl-acetamide, n-methoxy-n-methylacetamid, pubchem12775, acmc-209pdo, n-methoxy-n-methylacetamde, n-methoxy n-methylacetamide, n-methoxy-n-methyl acetamide
PubChem CID 537505
IUPAC Name N-methoxy-N-methylacetamide
SMILES CON(C)C(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9700g/mL
Boiling Point 152°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4240 to 1.426
Linear Formula CH3CON(OCH3)CH3
Quantity 5 g
Specific Gravity 0.97
Solubility Information Solubility in water: soluble
Formula Weight 103.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Methoxy-N-methylacetamide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only