missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methyl-2-furancarboxamide 98.0+%, TCI America™

Catalog No. M14761G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14761G Supplier TCI America Supplier No. M14761G
Only null left

Chemical Identifiers

CAS 95091-92-2
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.153
InChI Key ZPMBEUDMVUSUOS-UHFFFAOYSA-N
Synonym N-Methoxy-N-methyl-2-furamide
PubChem CID 10820793
IUPAC Name N-methoxy-N-methylfuran-2-carboxamide
SMILES CN(C(=O)C1=CC=CO1)OC

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 155.15
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material N-Methoxy-N-methyl-2-furancarboxamide
TSCA No
Recommended Storage Refrigerator