missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine sodium salt, ultrapure, 97%, Thermo Scientific Chemicals

$144.04 - $144.04

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15H28NNaO3
Molecular Weight (g/mol) 293.38
MDL Number MFCD00042728
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
Synonym sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
PubChem CID 23668817
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6492818
View Documents
Thermo Scientific Chemicals
J6492818
50 g
Each for $144.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

137-16-6
293.38
KSAVQLQVUXSOCR-UHFFFAOYSA-M
23668817
C15H28NNaO3
MFCD00042728
sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

137-16-6
White
MFCD00042728
5322974
14,4368
Soluble in water
[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
293.38
293.39
Powder
Ultrapure
44°C to 47°C
C15H28NNaO3
50 g
Hygroscopic
sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
KSAVQLQVUXSOCR-UHFFFAOYSA-M
sodium;2-[dodecanoyl(methyl)amino]acetate
23668817
97%
N-Lauroylsarcosine sodium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P332+P313-P362-P501c

H315-H318-H330

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 205-281-5

RTECSNumber : MC0598960

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only