missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine sodium salt, 95%

Catalog No. p-4228757
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434370250 25 g Glass bottle
AC434371000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434370250 Supplier Thermo Scientific Chemicals Supplier No. 434370250
Only null left

CAS: 137-16-6 | C15H28NNaO3 | 293.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15H28NNaO3
Molecular Weight (g/mol) 293.38
MDL Number MFCD00042728
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
Synonym sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
PubChem CID 23668817
IUPAC Name sodium;2-[dodecanoyl(methyl)amino]acetate
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

CAS Min % 94
CAS Max % 100
Melting Point 45.0°C to 47.0°C
Color White
pH 7.0 to 9.0
Loss on Drying 7% max. ()
Infrared Spectrum Authentic
Assay Percent Range 0.95
Packaging Glass bottle
Linear Formula NaCO2CH2N(CH3)CO(CH2)10CH3
Quantity 25 g
Merck Index 154401
Solubility Information (10% in water) Almost clear colorless to light yellow
Formula Weight 293.39
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Lauroylsarcosine sodium salt
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 1), Acute toxicity (category 2), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye damage, Fatal if inhaled, Causes skin irritation
EINECSNumber 205-281-5

RUO – Research Use Only