Learn More
N-Lauroylsarcosine sodium salt, 95%
CAS: 137-16-6 | C15H28NNaO3 | 293.38 g/mol
Supplier: Thermo Scientific Chemicals J6004018
Description
N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoprotein's; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungi static agent in aqueous dispersion (1%).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoprotein′s; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungi static agent in aqueous dispersion (1%).
Solubility
Soluble in water (293 g/L).
Notes
Hygroscopic, store away form water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent and reducing agent.
Specifications
| N-Lauroylsarcosine sodium salt | |
| 44°C to 47°C | |
| C15H28NNaO3 | |
| MFCD00042728 | |
| 5322974 | |
| 14,4368 | |
| Soluble in water (293g/L). | |
| [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O | |
| 293.38 | |
| 293.38 | |
| Crystalline Powder |
| 137-16-6 | |
| White | |
| CH3(CH2)10CON(CH3)CH2COONa | |
| 50 g | |
| Hygroscopic | |
| sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30 | |
| KSAVQLQVUXSOCR-UHFFFAOYSA-M | |
| sodium;2-[dodecanoyl(methyl)amino]acetate | |
| 23668817 | |
| 95% |
Chemical Identifiers
| 137-16-6 | |
| 293.38 | |
| KSAVQLQVUXSOCR-UHFFFAOYSA-M | |
| 23668817 | |
| [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O |
| C15H28NNaO3 | |
| MFCD00042728 | |
| sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30 | |
| sodium;2-[dodecanoyl(methyl)amino]acetate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P332+P313-P362-P501c
H315-H318-H330
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 205-281-5
RTECSNumber : MC0598960
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only