missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine sodium salt, 95%

Catalog No. aaj6004009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6004009 10 g
AAJ6004018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6004009 Supplier Thermo Scientific Chemicals Supplier No. J6004009
Only null left

CAS: 137-16-6 | C15H28NNaO3 | 293.38 g/mol

N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoprotein's; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungi static agent in aqueous dispersion (1%).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoprotein′s; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungi static agent in aqueous dispersion (1%).

Solubility
Soluble in water (293 g/L).

Notes
Hygroscopic, store away form water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent and reducing agent.

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15H28NNaO3
Molecular Weight (g/mol) 293.38
MDL Number MFCD00042728
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
Synonym sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
PubChem CID 23668817
IUPAC Name sodium;2-[dodecanoyl(methyl)amino]acetate
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

Melting Point 44°C to 47°C
Color White
Linear Formula CH3(CH2)10CON(CH3)CH2COONa
Quantity 10 g
Beilstein 5322974
Sensitivity Hygroscopic
Merck Index 14,4368
Solubility Information Soluble in water (293g/L).
Formula Weight 293.38
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material N-Lauroylsarcosine sodium salt
Show More Show Less
Hazard Category H315-H318-H330
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 205-281-5
RTECSNumber MC0598960
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only