missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine, 98%, MP Biomedicals™

Catalog No. ICN19028990
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19028990 500 g
ICN19028980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19028990 Supplier MP Biomedicals Inc Supplier No. 0219028990

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15H28NNaO3
Molecular Weight (g/mol) 293.38
MDL Number MFCD00042728
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
Synonym sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
PubChem CID 23668817
IUPAC Name sodium;2-[dodecanoyl(methyl)amino]acetate
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

Color White
Assay Percent Range 0.98
Quantity 500 g
Formula Weight 293.4
Physical Form Powder
Chemical Name or Material N-Lauroylsarcosine