missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Isovalerylglycine 98.0+%, TCI America™
$192.00 - $1014.05
Chemical Identifiers
| CAS | 16284-60-9 |
|---|---|
| Molecular Formula | C7H13NO3 |
| Molecular Weight (g/mol) | 159.19 |
| MDL Number | MFCD00059629 |
| InChI Key | ZRQXMKMBBMNNQC-UHFFFAOYSA-N |
| Synonym | N-Isovaleroylglycine |
| PubChem CID | 546304 |
| ChEBI | CHEBI:70984 |
| IUPAC Name | 2-(3-methylbutanamido)acetic acid |
| SMILES | CC(C)CC(=O)NCC(O)=O |
Chemical Identifiers
| 16284-60-9 | |
| 159.19 | |
| ZRQXMKMBBMNNQC-UHFFFAOYSA-N | |
| 546304 | |
| 2-(3-methylbutanamido)acetic acid |
| C7H13NO3 | |
| MFCD00059629 | |
| N-Isovaleroylglycine | |
| CHEBI:70984 | |
| CC(C)CC(=O)NCC(O)=O |
Specifications
| 16284-60-9 | |
| White-Yellow | |
| MFCD00059629 | |
| N-Isovaleroylglycine | |
| CC(C)CC(=O)NCC(O)=O | |
| 159.19 | |
| CHEBI:70984 | |
| ≥98.0% (T) | |
| N-Isovalerylglycine |
| 105°C | |
| C7H13NO3 | |
| 1 g | |
| ZRQXMKMBBMNNQC-UHFFFAOYSA-N | |
| 2-(3-methylbutanamido)acetic acid | |
| 546304 | |
| 159.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No