missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylmethylamine, 98%

Catalog No. p-7056772
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6589206 5 g
AAH6589214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6589206 Supplier Thermo Scientific Chemicals Supplier No. H6589206

CAS: 4747-21-1 | C4H11N | 73.139 g/mol

N-Isopropylmethylamine is used in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (RAFT) polymerization. It is suitable for use in the synthesis of novel 5-ethoxy-N,N-dialkyl-[α,α,β,β-H4]-tryptamines and their [α,α,β,β,β-D4]-counterparts following the Speeter and Anthony procedure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isopropylmethylamine is used in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (RAFT) polymerization. It is suitable for use in the synthesis of novel 5-ethoxy-N,N-dialkyl-[α,α,β,β-H4]-tryptamines and their [α,α,β,β,β-D4]-counterparts following the Speeter and Anthony procedure.

Solubility
Soluble in chloroform, DMSO, methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Do not store together with acids. Incompatible with acids , air and oxidizing agents.

Chemical Identifiers

CAS 4747-21-1
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00042859
InChI Key XHFGWHUWQXTGAT-UHFFFAOYSA-N
Synonym n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine
PubChem CID 78485
IUPAC Name N-methylpropan-2-amine
SMILES CC(C)NC

Specifications

Color Colorless
Density 0.702 g/mL
Boiling Point 50°C to 53°C
Flash Point -32°C (-26°F)
Refractive Index 1.348
Linear Formula (CH3)2CHNHCH3
Quantity 5 g
UN Number UN2734
Beilstein 1730877
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,DMSO,methanol.
Formula Weight 73.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Isopropylmethylamine
Show More Show Less
Hazard Category H225-H301+H331-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: I; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 225-266-7
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only