missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Isopropylaniline 99.0+%, TCI America™
$102.52 - $507.02
Chemical Identifiers
| CAS | 768-52-5 |
|---|---|
| Molecular Formula | C9H13N |
| Molecular Weight (g/mol) | 135.21 |
| MDL Number | MFCD00026347 |
| InChI Key | FRCFWPVMFJMNDP-UHFFFAOYSA-N |
| Synonym | n-isopropylaniline, isopropylaniline, n-phenylisopropylamine, benzenamine, n-1-methylethyl, aniline, n-isopropyl, n-propan-2-yl aniline, n-iso-propylaniline, unii-i7y9616f3r, ccris 4831, 2-phenylamino propane |
| PubChem CID | 13032 |
| IUPAC Name | N-propan-2-ylaniline |
| SMILES | CC(C)NC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I038425ML
|
TCI America
I038425ML |
25 mL |
Each for $102.52
|
|
|||||
|
I0384100ML
|
TCI America
I0384100ML |
100 mL |
Each for $219.95
|
|
|||||
|
I0384500ML
|
TCI America
I0384500ML |
500 mL |
Each for $507.02
|
|
|||||
Chemical Identifiers
| 768-52-5 | |
| 135.21 | |
| FRCFWPVMFJMNDP-UHFFFAOYSA-N | |
| 13032 | |
| CC(C)NC1=CC=CC=C1 |
| C9H13N | |
| MFCD00026347 | |
| n-isopropylaniline, isopropylaniline, n-phenylisopropylamine, benzenamine, n-1-methylethyl, aniline, n-isopropyl, n-propan-2-yl aniline, n-iso-propylaniline, unii-i7y9616f3r, ccris 4831, 2-phenylamino propane | |
| N-propan-2-ylaniline |
Specifications
| 768-52-5 | |
| Yellow | |
| C9H13N | |
| 25 mL | |
| FRCFWPVMFJMNDP-UHFFFAOYSA-N | |
| N-propan-2-ylaniline | |
| 13032 | |
| ≥99.0% (GC,T) | |
| N-Isopropylaniline |
| -50°C | |
| 213°C | |
| MFCD00026347 | |
| n-isopropylaniline, isopropylaniline, n-phenylisopropylamine, benzenamine, n-1-methylethyl, aniline, n-isopropyl, n-propan-2-yl aniline, n-iso-propylaniline, unii-i7y9616f3r, ccris 4831, 2-phenylamino propane | |
| CC(C)NC1=CC=CC=C1 | |
| 135.21 | |
| 135.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0118&(3)-0206
RTECSNumber : BY4190000
TSCA : Yes