missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylaniline 99.0+%, TCI America™

Catalog No. I0384500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0384500ML 500 mL
I038425ML 25 mL
I0384100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0384500ML Supplier TCI America Supplier No. I0384500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 768-52-5
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00026347
InChI Key FRCFWPVMFJMNDP-UHFFFAOYSA-N
Synonym n-isopropylaniline, isopropylaniline, n-phenylisopropylamine, benzenamine, n-1-methylethyl, aniline, n-isopropyl, n-propan-2-yl aniline, n-iso-propylaniline, unii-i7y9616f3r, ccris 4831, 2-phenylamino propane
PubChem CID 13032
IUPAC Name N-propan-2-ylaniline
SMILES CC(C)NC1=CC=CC=C1

Specifications

Melting Point -50°C
Color Yellow
Boiling Point 213°C
Quantity 500 mL
Formula Weight 135.21
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Isopropylaniline
EINECSNumber (3)-0118&(3)-0206
RTECSNumber BY4190000
TSCA Yes