missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Isopropylaniline 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | -50°C |
| Color | Yellow |
| Boiling Point | 213°C |
| Quantity | 100 mL |
| Formula Weight | 135.21 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | N-Isopropylaniline |
Safety and Handling
| EINECSNumber | (3)-0118&(3)-0206 |
| RTECSNumber | BY4190000 |
| TSCA | Yes |