missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylaniline, 98%

CAS: 768-52-5 | C9H13N | 135.21 g/mol

$92.51 - $278.92

Chemical Identifiers

CAS 768-52-5
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00026347
InChI Key FRCFWPVMFJMNDP-UHFFFAOYSA-N
Synonym n-isopropylaniline, isopropylaniline, n-phenylisopropylamine, benzenamine, n-1-methylethyl, aniline, n-isopropyl, n-propan-2-yl aniline, n-iso-propylaniline, unii-i7y9616f3r, ccris 4831, 2-phenylamino propane
PubChem CID 13032
IUPAC Name N-propan-2-ylaniline
SMILES CC(C)NC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1064609
View Documents
Thermo Scientific Chemicals
L1064609
10 g
Each for $92.51
Only null left
 
AAL1064618
View Documents
Thermo Scientific Chemicals
L1064618
50 g
Each for $278.92
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

768-52-5
135.21
FRCFWPVMFJMNDP-UHFFFAOYSA-N
13032
CC(C)NC1=CC=CC=C1
C9H13N
MFCD00026347
n-isopropylaniline, isopropylaniline, n-phenylisopropylamine, benzenamine, n-1-methylethyl, aniline, n-isopropyl, n-propan-2-yl aniline, n-iso-propylaniline, unii-i7y9616f3r, ccris 4831, 2-phenylamino propane
N-propan-2-ylaniline

Specifications

768-52-5
0.938
88°C (190°F)
1.5385
10 g
2205871
FRCFWPVMFJMNDP-UHFFFAOYSA-N
N-propan-2-ylaniline
13032
98%
-32°C
203°C to 204°C
C9H13N
MFCD00026347
UN2810
n-isopropylaniline, isopropylaniline, n-phenylisopropylamine, benzenamine, n-1-methylethyl, aniline, n-isopropyl, n-propan-2-yl aniline, n-iso-propylaniline, unii-i7y9616f3r, ccris 4831, 2-phenylamino propane
CC(C)NC1=CC=CC=C1
135.21
135.21
N-Isopropylaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H302
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.

P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319-H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 212-196-7

RTECSNumber : BY4190000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only