missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylaniline, 98%

Catalog No. AAL1064609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1064609 10 g
AAL1064618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1064609 Supplier Thermo Scientific Chemicals Supplier No. L1064609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 768-52-5 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 768-52-5
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00026347
InChI Key FRCFWPVMFJMNDP-UHFFFAOYSA-N
Synonym n-isopropylaniline, isopropylaniline, n-phenylisopropylamine, benzenamine, n-1-methylethyl, aniline, n-isopropyl, n-propan-2-yl aniline, n-iso-propylaniline, unii-i7y9616f3r, ccris 4831, 2-phenylamino propane
PubChem CID 13032
IUPAC Name N-propan-2-ylaniline
SMILES CC(C)NC1=CC=CC=C1

Specifications

Melting Point -32°C
Density 0.938
Boiling Point 203°C to 204°C
Flash Point 88°C (190°F)
Refractive Index 1.5385
Quantity 10 g
UN Number UN2810
Beilstein 2205871
Formula Weight 135.21
Percent Purity 98%
Chemical Name or Material N-Isopropylaniline
Show More Show Less
Hazard Category H227-H302-H315-H319-H330
Hazard Statement GHS H Statement
H331-H227-H302
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 212-196-7
RTECSNumber BY4190000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only