missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylacrylamide (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. I040125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I040125G 25 g
I0401100G 100 g
I0401500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I040125G Supplier TCI America Supplier No. I040125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2210-25-5
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00041913
InChI Key QNILTEGFHQSKFF-UHFFFAOYSA-N
Synonym n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u
PubChem CID 16637
IUPAC Name N-(propan-2-yl)prop-2-enamide
SMILES CC(C)NC(=O)C=C

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 110°C
Quantity 25 g
Formula Weight 113.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Isopropylacrylamide (stabilized with MEHQ)
EINECSNumber (2)-1015
RTECSNumber AS3675000
TSCA Yes
Recommended Storage Refrigerator