missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Isopropylacrylamide, 99%, pure, stabilized

Catalog No. AC412780250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC412780250 25 g
AC412781000 100 g
AC412785000 500 g
AC412780025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC412780250 Supplier Thermo Scientific Chemicals Supplier No. 412780250

CAS: 2210-25-5 | C6H11NO | 113.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2210-25-5
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00041913
InChI Key QNILTEGFHQSKFF-UHFFFAOYSA-N
Synonym n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u
PubChem CID 16637
IUPAC Name N-propan-2-ylprop-2-enamide
SMILES CC(C)NC(=O)C=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 62.0°C to 66.0°C
Color White to Yellow
Boiling Point 89.0°C to 92.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH2CHCONHCH(CH3)2
Quantity 25 g
Solubility Information Solubility in water: soluble
Stabilizer 500ppm (ca.) MEHQ
Formula Weight 113.16
Percent Purity 99%
Grade Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N-Isopropylacrylamide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-638-5
RTECSNumber AS3675000
TSCA TSCA

RUO – Research Use Only