missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylacrylamide, 97%

Catalog No. AAH6626222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6626222 100 g
AAH6626214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6626222 Supplier Thermo Scientific Chemicals Supplier No. H6626222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2210-25-5 | C6H11NO | 113.16 g/mol

N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA, pNIPAAm, pNIPAA or pNIPAm) thermo sensitive polymer- or copolymer-based hydrogels. Polymers that contain NIPAM shrink dramatically above 33°C. Monomer used in the preparation of thermally sensitive, water-swellable hydrogels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA, pNIPAAm, pNIPAA or pNIPAm) thermo sensitive polymer- or copolymer-based hydrogels. Polymers that contain NIPAM shrink dramatically above 33°C. Monomer used in the preparation of thermally sensitive, water-swellable hydrogels.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 2210-25-5
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00041913
InChI Key QNILTEGFHQSKFF-UHFFFAOYSA-N
Synonym n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u
PubChem CID 16637
SMILES CC(C)NC(=O)C=C

Specifications

Melting Point 60°C to 63°C
Boiling Point 89°C to 92°C (2 mmHg)
Quantity 100 g
Beilstein 1744926
Solubility Information Slightly soluble in water.
IUPAC Name N-propan-2-ylprop-2-enamide
Formula Weight 113.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Isopropylacrylamide
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 218-638-5
RTECSNumber AS3675000
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only