missing translation for 'onlineSavingsMsg'
Learn More

N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%, Thermo Scientific™

Catalog No. p-7035024
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5272603 1g
AAH5272606 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5272603 Supplier Thermo Scientific Chemicals Supplier No. H5272603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Isobutyryl-5′-O-tert-butyldimethylsilyl-2′-deoxyguanosine is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Store away from oxidizing agents.

Chemical Identifiers

CAS 85326-10-9
Molecular Formula C20H33N5O5Si
Molecular Weight (g/mol) 451.599
MDL Number MFCD04972287
InChI Key WZQWRHWLQPEBFF-BFHYXJOUSA-N
Synonym n2-isobutyryl-5'-o-tert-butyldimethylsilyl-2'-deoxyguanosine, n-9-2r,4s,5r-5-tert-butyldimethylsilyl oxy methyl-4-hydroxyoxolan-2-yl-6-oxo-1h-purin-2-yl-2-methylpropanamide
PubChem CID 12016034
IUPAC Name N-[9-[(2R,4S,5R)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide
SMILES CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C3CC(C(O3)CO[Si](C)(C)C(C)(C)C)O

Specifications

Quantity 1g
Sensitivity Moisture sensitive
Formula Weight 451.6
Percent Purity 97%
Chemical Name or Material N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only