missing translation for 'onlineSavingsMsg'
Learn More

N-Iodosuccinimide, 98%

Catalog No. ac299571000
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC299571000 100 g Glass bottle
AC299570100 10 g Glass bottle
AC299570500 50 g Glass bottle
AC299575000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC299571000 Supplier Thermo Scientific Chemicals Supplier No. 299571000
Only null left

CAS: 516-12-1 | C4H4INO2 | 224.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 516-12-1
Molecular Formula C4H4INO2
Molecular Weight (g/mol) 224.99
MDL Number MFCD00005512
InChI Key LQZMLBORDGWNPD-UHFFFAOYSA-N
Synonym n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione
PubChem CID 120273
ChEBI CHEBI:53204
IUPAC Name 1-iodopyrrolidine-2,5-dione
SMILES IN1C(=O)CCC1=O

Specifications

Melting Point 197.0°C to 204.0°C
Color Brown to White-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Quantity 100 g
Merck Index 15,5088
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in acetone,methanol,moderately soluble in dioxane,slightly soluble in methylene chloride and ethyl a,practically insoluble in carbon tetrachloride,eth
Formula Weight 224.99
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Iodosuccinimide
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 208-221-6

RUO – Research Use Only