missing translation for 'onlineSavingsMsg'
Learn More

N-(Imidazo[1,2-a]pyridin-6-ylmethyl)-N-methylamine, ≥97%, Thermo Scientific™

Catalog No. CC33146CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC33146CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC33146CB Supplier Thermo Scientific Chemicals Supplier No. CC33146CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 864068-82-6
Molecular Formula C9H11N3
Molecular Weight (g/mol) 161.21
MDL Number MFCD08271888
InChI Key VTAYGJSBPFKWJX-UHFFFAOYSA-N
Synonym n-imidazo 1,2-a pyridin-6-ylmethyl-n-methylamine, imidazo 1,2-a pyridin-6-ylmethyl methyl amine, 1-imidazo 1,2-a pyridin-6-yl-n-methylmethanamine, 4-hydroimidazo 1,2-a pyridin-6-ylmethyl methylamine, imidazo 1,2-a pyridine-6-methanamine,n-methyl
PubChem CID 7537497
IUPAC Name 1-imidazo[1,2-a]pyridin-6-yl-N-methylmethanamine
SMILES CNCC1=CN2C=CN=C2C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 161.21
Percent Purity 95%
Chemical Name or Material N-(Imidazo[1, 2-a]pyridin-6-ylmethyl)-N-methylamine
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.