missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich N-Hydroxysulfosuccinimide Sodium Salt
Description
N-Hydroxysulfosuccinimide sodium salt is a water-soluble sulfonated analog of N-hydroxysuccinimide. It can undergo a coupling reaction with carboxylic acids in the presence of carbodiimides such as 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide(EDC) to form hydrophilic N-hydroxysulfosuccinimide active esters. These esters are useful in protein cross-linking experiments.Sulfo-NHS is used for preparing hydrophilic active esters, e.g. for crosslinking agents
Specifications
Specifications
| CAS | 106627-54-7 |
| Molecular Formula | C4H4NNaO6S |
| Linear Formula | C4H4NNaO6S |
| MDL Number | MFCD00043100 |
| Quantity | 1 g |
| Synonym | Hydroxy-2,5-dioxopyrrolidine-3-sulfonicacid sodium salt; Sulfo-NHS |
| Molecular Weight (g/mol) | 217.13 g/mol |