missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxysuccinimide 98.0+%, TCI America™

Catalog No. h062325g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
H062325G 25 g
H0623100G 100 g
H0623500G 500 g
3 options

Catalog No. H062325G

Supplier: TCI America H062325G

Only null left

Chemical Identifiers

CAS 6066-82-6
Molecular Formula C4H5NO3
Molecular Weight (g/mol) 115.088
MDL Number MFCD00005516
InChI Key NQTADLQHYWFPDB-UHFFFAOYSA-N
Synonym n-hydroxysuccinimide, hosu, 1-hydroxysuccinimide, 2,5-pyrrolidinedione, 1-hydroxy, 1-hydroxy-2,5-pyrrolidinedione, succinimide, n-hydroxy, n-hydroxysuccinimde, n-hydroxysuccinimid, unii-mje3791m4t, ccris 2604
PubChem CID 80170
IUPAC Name 1-hydroxypyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)O

Specifications

Melting Point 98°C
Color White
Quantity 25 g
Formula Weight 115.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Hydroxysuccinimide
EINECSNumber (5)-5911
TSCA Yes