missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxysuccinimide, 98+%

Catalog No. AC157270250
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157270250 25 g Plastic bottle
AC157271000 100 g Plastic bottle
AC157272500 250 g Plastic bottle
AC157270010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC157270250 Supplier Thermo Scientific Chemicals Supplier No. 157270250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6066-82-6 | C4H5NO3 | 115.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 6066-82-6
Molecular Formula C4H5NO3
Molecular Weight (g/mol) 115.09
MDL Number MFCD00005516
InChI Key NQTADLQHYWFPDB-UHFFFAOYSA-N
Synonym n-hydroxysuccinimide, hosu, 1-hydroxysuccinimide, 2,5-pyrrolidinedione, 1-hydroxy, 1-hydroxy-2,5-pyrrolidinedione, succinimide, n-hydroxy, n-hydroxysuccinimde, n-hydroxysuccinimid, unii-mje3791m4t, ccris 2604
PubChem CID 80170
IUPAC Name 1-hydroxypyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 93.0°C to 95.0°C
Color Brown to Brown-Yellow
Loss on Drying 0.5% max. (60°C, 2 hrs, 1mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Quantity 25 g
Beilstein 21, 380
Fieser 01,487; 09,246
Merck Index 15, 4884
Solubility Information which is virtually insoluble in acetone
Formula Weight 115.09
Percent Purity 98+%
Physical Form Liquid or Solid
Chemical Name or Material N-Hydroxysuccinimide, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-001-3
TSCA TSCA
Recommended Storage This product contains some hydroxysuccinimide:ammonia adduct, which is virtually insoluble in acetone. The product has a tendency to caking.

RUO – Research Use Only