Learn More
N-(Hydroxymethyl)trifluoroacetamide, 98+%
CAS: 50667-69-1 | C3H4F3NO2 | 143.065 g/mol
$120.56 - $400.73
Chemical Identifiers
| CAS | 50667-69-1 |
|---|---|
| Molecular Formula | C3H4F3NO2 |
| Molecular Weight (g/mol) | 143.065 |
| MDL Number | MFCD00014416 |
| InChI Key | URUWXKFAEKTWKG-UHFFFAOYSA-N |
| Synonym | 2,2,2-trifluoro-n-hydroxymethyl acetamide, n-hydroxymethyl trifluoroacetamide, n-hydroxymethyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-hydroxymethyl, acmc-20akaf, trifluoroacetamidomethanol, n-hydroxymethyl fluoroacetamide, n-hydroxymethyl-trifluoroacetamide, n-hydroxy methyl trifluoro acetamide |
| PubChem CID | 3084931 |
| IUPAC Name | 2,2,2-trifluoro-N-(hydroxymethyl)acetamide |
| SMILES | C(NC(=O)C(F)(F)F)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0012806
|
Thermo Scientific Chemicals
L0012806 |
5 g |
Each for $120.56
|
|
|||||
|
AAL0012814
|
Thermo Scientific Chemicals
L0012814 |
25 g |
Each for $400.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 50667-69-1 | |
| 143.065 | |
| URUWXKFAEKTWKG-UHFFFAOYSA-N | |
| 3084931 | |
| C(NC(=O)C(F)(F)F)O |
| C3H4F3NO2 | |
| MFCD00014416 | |
| 2,2,2-trifluoro-n-hydroxymethyl acetamide, n-hydroxymethyl trifluoroacetamide, n-hydroxymethyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-hydroxymethyl, acmc-20akaf, trifluoroacetamidomethanol, n-hydroxymethyl fluoroacetamide, n-hydroxymethyl-trifluoroacetamide, n-hydroxy methyl trifluoro acetamide | |
| 2,2,2-trifluoro-N-(hydroxymethyl)acetamide |
Specifications
| 50667-69-1 | |
| C3H4F3NO2 | |
| MFCD00014416 | |
| 1859914 | |
| 2,2,2-trifluoro-n-hydroxymethyl acetamide, n-hydroxymethyl trifluoroacetamide, n-hydroxymethyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-hydroxymethyl, acmc-20akaf, trifluoroacetamidomethanol, n-hydroxymethyl fluoroacetamide, n-hydroxymethyl-trifluoroacetamide, n-hydroxy methyl trifluoro acetamide | |
| C(NC(=O)C(F)(F)F)O | |
| 143.065 | |
| 143.06 | |
| N-(Hydroxymethyl)trifluoroacetamide |
| 99°C to 103°C | |
| CF3CONHCH2OH | |
| 5 g | |
| Hygroscopic | |
| URUWXKFAEKTWKG-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-N-(hydroxymethyl)acetamide | |
| 3084931 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 256-698-4
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only