missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Hydroxymethyl)nicotinamide 97.0+%, TCI America™
$74.56 - $713.94
Chemical Identifiers
| CAS | 3569-99-1 |
|---|---|
| Molecular Formula | C7H8N2O2 |
| Molecular Weight (g/mol) | 152.153 |
| MDL Number | MFCD00010437 |
| InChI Key | JRFKIOFLCXKVOT-UHFFFAOYSA-N |
| PubChem CID | 77116 |
| IUPAC Name | N-(hydroxymethyl)pyridine-3-carboxamide |
| SMILES | C1=CC(=CN=C1)C(=O)NCO |
Chemical Identifiers
| 3569-99-1 | |
| 152.153 | |
| JRFKIOFLCXKVOT-UHFFFAOYSA-N | |
| N-(hydroxymethyl)pyridine-3-carboxamide |
| C7H8N2O2 | |
| MFCD00010437 | |
| 77116 | |
| C1=CC(=CN=C1)C(=O)NCO |
Specifications
| 3569-99-1 | |
| White | |
| MFCD00010437 | |
| JRFKIOFLCXKVOT-UHFFFAOYSA-N | |
| N-(hydroxymethyl)pyridine-3-carboxamide | |
| 77116 | |
| ≥97.0% (HPLC,T) | |
| N-(Hydroxymethyl)nicotinamide |
| 145°C | |
| C7H8N2O2 | |
| 25 g | |
| C1=CC(=CN=C1)C(=O)NCO | |
| 152.153 | |
| 152.15 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : QS4476500
TSCA : No