missing translation for 'onlineSavingsMsg'
Learn More

N-(Hydroxymethyl)benzamide, 98%

Catalog No. AAA1225030
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1225030 250 g
AAA1225009 10 g
AAA1225014 25 g
AAA1225022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1225030 Supplier Thermo Scientific Chemicals Supplier No. A1225030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6282-02-6 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6282-02-6
MDL Number MFCD00014415

Specifications

Melting Point 94°C to 96°C
Molecular Formula C8H9NO2
Quantity 250 g
Beilstein 2045129
Synonym n-hydroxymethyl benzamide, benzamide, n-hydroxymethyl, n-methylolbenzamide, acmc-20akah, pubchem12540, enamine_001590, n-hydroxymetyl-benzamide, benzamide,n-hydroxymethyl, bis 5-nitropyridin-4-yl sulfane
InChI Key UOUBPDZUBVJZOQ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)NCO
IUPAC Name N-(hydroxymethyl)benzamide
Molecular Weight (g/mol) 151.17
PubChem CID 80493
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material N- (Hydroxymethyl)benzamide
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 228-495-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only