missing translation for 'onlineSavingsMsg'
Learn More

N-(Hydroxymethyl)acrylamide 98.0+%, TCI America™

Catalog No. p-7132246
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M057425G 25 g
M0574500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M057425G Supplier TCI America Supplier No. M057425G

Chemical Identifiers

CAS 924-42-5
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.105
MDL Number MFCD00004597
InChI Key CNCOEDDPFOAUMB-UHFFFAOYSA-N
Synonym n-methylolacrylamide, n-hydroxymethyl acrylamide, n-methanolacrylamide, monomethylolacrylamide, methylolacrylamide, n-methylol acrylamide, 2-propenamide, n-hydroxymethyl, acrylamide, n-hydroxymethyl, uramine t 80, n-hydroxymethyl-2-propenamide
PubChem CID 13543
ChEBI CHEBI:82492
IUPAC Name N-(hydroxymethyl)prop-2-enamide
SMILES C=CC(=O)NCO

Specifications

Melting Point 76°C
Color White
Quantity 25 g
UN Number 3531
Formula Weight 101.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(Hydroxymethyl)acrylamide
EINECSNumber (2)-1022
RTECSNumber AS3600000
TSCA Yes
Recommended Storage Refrigerator