missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Hydroxymethyl)acetamide, 98%
CAS: 625-51-4 | C3H7NO2 | 89.09 g/mol
Supplier: Thermo Scientific Chemicals A1382014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-(Hydroxymethyl)acetamide | |
| 47°C to 52°C | |
| C3H7NO2 | |
| 25 g | |
| Hygroscopic | |
| n-hydroxymethyl acetamide, acetamidomethanol, formicin, acetamide, n-hydroxymethyl, unii-rcg03a51mo, n-hydroxymethyl-acetamide, rcg03a51mo, acetamido methanol, n-acetylmethanolamine, acetylamino methanol | |
| CC(=O)NCO | |
| 89.09 | |
| 89.09 |
| 625-51-4 | |
| 1.14 | |
| MFCD00014417 | |
| 506226 | |
| 14,4242 | |
| HWJHZLJIIWOTGZ-UHFFFAOYSA-N | |
| N-(hydroxymethyl)acetamide | |
| 69365 | |
| 98% |
Chemical Identifiers
| 625-51-4 | |
| 89.09 | |
| HWJHZLJIIWOTGZ-UHFFFAOYSA-N | |
| 69365 | |
| CC(=O)NCO |
| C3H7NO2 | |
| MFCD00014417 | |
| n-hydroxymethyl acetamide, acetamidomethanol, formicin, acetamide, n-hydroxymethyl, unii-rcg03a51mo, n-hydroxymethyl-acetamide, rcg03a51mo, acetamido methanol, n-acetylmethanolamine, acetylamino methanol | |
| N-(hydroxymethyl)acetamide |
Safety and Handling
EINECSNumber : 210-897-2
RTECSNumber : AC3610000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only