missing translation for 'onlineSavingsMsg'
Learn More

N-(Hydroxymethyl)acetamide, 98%

Catalog No. AAA1382009
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1382009 10 g
AAA1382014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1382009 Supplier Thermo Scientific Chemicals Supplier No. A1382009

CAS: 625-51-4 | C3H7NO2 | 89.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 625-51-4
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00014417
InChI Key HWJHZLJIIWOTGZ-UHFFFAOYSA-N
Synonym n-hydroxymethyl acetamide, acetamidomethanol, formicin, acetamide, n-hydroxymethyl, unii-rcg03a51mo, n-hydroxymethyl-acetamide, rcg03a51mo, acetamido methanol, n-acetylmethanolamine, acetylamino methanol
PubChem CID 69365
IUPAC Name N-(hydroxymethyl)acetamide
SMILES CC(=O)NCO

Specifications

Melting Point 47°C to 52°C
Density 1.14
Quantity 10 g
Beilstein 506226
Sensitivity Hygroscopic
Merck Index 14,4242
Formula Weight 89.09
Percent Purity 98%
Chemical Name or Material N-(Hydroxymethyl)acetamide
EINECSNumber 210-897-2
RTECSNumber AC3610000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only