missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy-5-norbornene-2,3-dicarboximide 99.0+%, TCI America™

Catalog No. H052825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H052825G 25 g
H0528250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H052825G Supplier TCI America Supplier No. H052825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21715-90-2
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.18
MDL Number MFCD00065691
InChI Key ZUSSTQCWRDLYJA-UHFFFAOYNA-N
Synonym honb, n-hydroxy-5-norbornene-2,3-dicarboximide, 2-hydroxy-3a,4,7,7a-tetrahydro-1h-4,7-methanoisoindole-1,3 2h-dione, 4-hydroxy-4-azatricyclo 5.2.1.0 2,6 dec-8-ene-3,5-dione, n-hydroxynorborn-5-ene-2,3-dicarboximide, n-hydroxy-5-norbornene-2,3-dicarboxylic imide, 5-norbornene-2, n-hydroxy, 4,7-methano-1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-2-hydroxy, 4,3 2h-dione, 3a,4,7,7a-tetrahydro-2-hydroxy, 4-hydroxy-4-azatricyclo 5.2.1.0<2,6> dec-8-ene-3,5-dione
PubChem CID 89529
IUPAC Name 4-hydroxy-4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
SMILES ON1C(=O)C2C3CC(C=C3)C2C1=O

Specifications

Melting Point 167°C
Color White
Quantity 25 g
Formula Weight 179.18
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Hydroxy-5-norbornene-2,3-dicarboximide [for Peptide Synthesis]