missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Hydroxy-2,2-diméthylpropanimidamide, 95%
Description
Ce Thermo Scientific Chemicals produit de marque faisait à l’origine partie du portefeuille de produits d’Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit / article original d’Acros Organics ou la référence SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | N-Hydroxy-2, 2-dimethylpropanimidamide |
| CAS % minimum | 94.0 |
| CAS % maximal | 100.0 |
| Point de fusion | 108,6 °C à 113,7 °C |
| Couleur | Blanc à brun |
| Spectre infrarouge | Authentique |
| Fourchette de pourcentages d’essai | 94% min. (HPLC) |
| Emballage | Bouteille en verre |
| Quantité | 5 g |
| Poids de formule | 116.16 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
Utilisation uniquement de la recherche RUO –