missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy-2,2-dimethylpropanimidamide, 95%

Catalog No. AC439660010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439660010 1 g Glass bottle
AC439660050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439660010 Supplier Thermo Scientific Chemicals Supplier No. 439660010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 42956-75-2 | C5H12N2O | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42956-75-2
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.16
InChI Key CVOGFMYWFRFWEQ-UHFFFAOYSA-N
Synonym n-hydroxypivalimidamide, 1z-n'-hydroxy-2,2-dimethylpropanimidamide, n-hydroxy-2,2-dimethylpropanimidamide, n-hydroxypivalamidine, z-n'-hydroxy-2,2-dimethylpropanimidamide, hydroxydimethylpropanimidamide, z-n'-hydroxypivalimidamide, z-n'-hydroxy-2,2-dimethylpropimidamide
PubChem CID 9582836
IUPAC Name N'-hydroxy-2,2-dimethylpropanimidamide
SMILES CC(C)(C)C(=NO)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 108.6°C to 113.7°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 116.16
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Hydroxy-2, 2-dimethylpropanimidamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only