missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyltriethoxysilane, 97%

Catalog No. p-7046838
Change view
Click to view available options
Quantity:
100 g
25 g
Catalog No. Quantity
AAL1652114 25 g
AAL1652122 100 g
2 options

Catalog No. AAL1652114

Supplier: Thermo Scientific Chemicals L1652114

Only null left

CAS: 18166-37-5 | C12H28O3Si | 248.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18166-37-5
Molecular Formula C12H28O3Si
Molecular Weight (g/mol) 248.44
MDL Number MFCD00087744
InChI Key WUMSTCDLAYQDNO-UHFFFAOYSA-N
Synonym hexyltriethoxysilane, triethoxy hexyl silane, hexyl triethoxysilane, n-hexyltriethoxysilane, hexyl-triethoxysilane, acmc-209ei3
PubChem CID 4437318
IUPAC Name triethoxy(hexyl)silane
SMILES CCCCCC[Si](OCC)(OCC)OCC

Specifications

Boiling Point 115°C (18mmHg)
Quantity 25 g
Linear Formula CH3(CH2)5Si(OCH2CH3)3
Beilstein 1757895
Flash Point 95°C (203°F)
Formula Weight 248.44
Percent Purity 97%
Sensitivity Moisture sensitive
Density 0.86
Chemical Name or Material n-Hexyltriethoxysilane
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only