missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyltriethoxysilane, 97%

Catalog No. AAL1652122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1652122 100 g
AAL1652114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1652122 Supplier Thermo Scientific Chemicals Supplier No. L1652122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18166-37-5 | C12H28O3Si | 248.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18166-37-5
Molecular Formula C12H28O3Si
Molecular Weight (g/mol) 248.44
MDL Number MFCD00087744
InChI Key WUMSTCDLAYQDNO-UHFFFAOYSA-N
Synonym hexyltriethoxysilane, triethoxy hexyl silane, hexyl triethoxysilane, n-hexyltriethoxysilane, hexyl-triethoxysilane, acmc-209ei3
PubChem CID 4437318
IUPAC Name triethoxy(hexyl)silane
SMILES CCCCCC[Si](OCC)(OCC)OCC

Specifications

Boiling Point 115°C (18mmHg)
Quantity 100 g
Flash Point 95°C (203°F)
Sensitivity Moisture sensitive
Density 0.86
Linear Formula CH3(CH2)5Si(OCH2CH3)3
Beilstein 1757895
Formula Weight 248.44
Percent Purity 97%
Chemical Name or Material n-Hexyltriethoxysilane
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only