missing translation for 'onlineSavingsMsg'
Learn More

n-Hexylbenzene, 98%

Catalog No. p-7020758
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1282114 25 g
AAA1282122 100 g
AAA1282136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1282114 Supplier Thermo Scientific Chemicals Supplier No. A1282114
Only null left

CAS: 1077-16-3 | C12H18 | 162.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1077-16-3
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00009526
InChI Key LTEQMZWBSYACLV-UHFFFAOYSA-N
Synonym 1-phenylhexane, n-hexylbenzene, benzene, hexyl, hexane, 1-phenyl, unii-8e3d1a2bzp, 8e3d1a2bzp, phenylhexane, hexyl-benzen, 1-hexylbenzene, 4-hexylbenzene
PubChem CID 14109
IUPAC Name hexylbenzene
SMILES CCCCCCC1=CC=CC=C1

Specifications

Melting Point -61°C
Density 0.861
Boiling Point 226°C to 227°C
Flash Point 83°C (181°F)
Refractive Index 1.486
Quantity 25 g
UN Number UN3082
Beilstein 1904786
Formula Weight 162.28
Percent Purity 98%
Chemical Name or Material n-Hexylbenzene
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 214-070-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only