missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyl acetate, 99%

Catalog No. AAL06342AK
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL06342AK 250 mL
AAL06342AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL06342AK Supplier Thermo Scientific Chemicals Supplier No. L06342AK
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 142-92-7 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
n-Hexyl acetate exhibits antimicrobial activity and can be used to improve the safety of minimally processed fruits. It is a fruity smelling fluid used as flavoring agent or in perfumes. It is a green leaf volatile from cabbage Brassica oleracea var. capitata L. It is used as plasticizers.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Store at 4°C. Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 142-92-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00009524
InChI Key AOGQPLXWSUTHQB-UHFFFAOYSA-N
Synonym n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural
PubChem CID 8908
ChEBI CHEBI:87510
IUPAC Name hexyl acetate
SMILES CCCCCCOC(=O)C

Specifications

Melting Point -80°C
Density 0.873
Boiling Point 167°C to 169°C
Flash Point 43°C (109°F)
Refractive Index 1.409
Linear Formula CH3CO2(CH2)5CH3
Quantity 250 mL
UN Number UN3271
Beilstein 1747138
Solubility Information Soluble in alcohol,ether. Insoluble in water.
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material n-Hexyl acetate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 205-572-7
RTECSNumber AI0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only